missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ StemPro™ Human Adipose-Derived Stem Cell Kit
StemPro™ Human Adipose-Derived Stem Cell Kit
Brand: Gibco™ R7788110
This item is not returnable.
View return policy
Description
Human adipose-derived stem cells (ADSCs) are isolated from human lipoaspirate tissue and cryopreserved from primary cultures. ADSCs have phenotypic and functional characteristics very similar to those of bone marrow-derived mesenchymal stem cells. Before cryopreservation, the ADSCs are expanded for one passage in MesenPRO RS™ Medium, a revolutionary new reduced-serum (2%) medium that reduces ADSC doubling times. The StemPro™ Human Adipose-Derived Stem Cell Kit contains cryopreserved normal human ADSCs and MesenPRO RS™ Medium - together, these components ensure optimal ADSC growth and expansion. Each lot of ADSCs originates from a single donor of human lipoaspirate tissue. Each vial of ADSCs contains cells that can differentiate into multiple mature cell phenotypes in vitro, including adipocytes, osteoblasts, and chondrocytes. ADSCs can be expanded to 4-5 passages before they lose their ability to grow or differentiate into all potential phenotypes. The ADSCs express the following flow cytometry cell-surface protein profile: positive CD29, CD44, CD73, CD90, CD105, CD166; negative CD14, CD31, CD45, Lin1.Applications:
Research use only. Not intended for human or animal diagnostic or therapeutic uses.
Note: MesenPRO RS™ Medium (Cat. no. 12746-012), composed of MesenPRO RS™ Growth Supplement and MesenPRO RS™ Basal Medium, is available for purchase separately.

Specifications
This kit is shipped in two parts with separate storage requirements. These components are not sold individually: –StemPro™ Human Adipose-Derived Stem Cells (one vial containing ∼1 x 106 cells; store in liquid nitrogen) and MesenPRO RS™ Growth Supplement (10 ml; store in the dark at -5 to -20°C) –MesenPRO RS™ Basal Medium (500 ml; store in the dark at 2-8°C) |
|
Human | |
Gibco™, StemPro™ |
Stem Cell (Mesenchymal) | |
Lipoaspirate | |
1 kit |
Product Suggestions
Customers who viewed this item also viewed
User manual for STEMPRO® Human Adipose-Derived Stem Cells
File size: 265.1 kB
User Guide: StemPro Human Adipose-Derived Stem Cells
Version: 2.0
File size: 229.8 kB
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Gibco™ StemPro™ Human Adipose-Derived Stem Cell Kit > 1 kit
For human ex vivo tissue and cell culture processing applications. CAUTION: When used as a medical device, Federal Law restricts this device to sale by or on the order of a physician.
Spot an opportunity for improvement?Share a Content Correction