missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Stericup™ E-GP Funnel-less Sterile Vacuum Filtration Systems
The newest addition to the Stericup™ Quick Release Family. Evolved with an eco-conscience.
Brand: Merck SEGPU1145
276.28 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
Stericup E and Steritop E sterile vacuum systems are the newest addition to the Stericup Quick Release Family. The Stericup E and Steritop E family of devices are designed as a more sustainable option for sterile filtration that significantly reduces plastic and packaging. These are funnel-less filter products for use in the sterile vacuum filtration of aqueous solutions such as media and tissue culture reagents/buffers.The Stericup E and Steritop E devices are designed specifically for sterile filtration from compatible bottles (38 and 45 mm neck size) of media and tissue culture reagents/buffers. Like Stericup Quick Release filters, Stericup E devices feature a tactile-stop cap to signal secure closure, an ample textured writing surface, and filter membrane characteristics clearly labeled on the filter collar.SEGPU0538 combines a 38 mm threaded collar, which uses a 0.22 μm pore size fast flow PES Express PLUS membrane, with a 500 mL receiver flask for sterilizing tissue culture media and buffers. Not recommended for reagents prepared in other vessel type devices like flasks or anything without a threaded neck.Specifications
Clear with blue adapter | |
Polystyrene | |
0.22 μm | |
40 cm² | |
45 mm threaded | |
Bottle Top Filter | |
73 mm |
Single-use | |
PES Express PLUS | |
Sterile | |
Sterile filtration | |
45 mm threaded | |
1000 mL | |
73 mm |