missing translation for 'onlineSavingsMsg'
Learn More

Contec™ Sterile Denatured Ethanol, 70%
BIOPHARMA

Blend of 70% v/v denatured ethanol with 30% purified water. Contec™ Sterile Low Endotoxin Alcohol, 70% is 0.2 micron filtered.

Brand:  Contec™ SBT170DEW

133.99 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.



 View more versions of this product

Product Code. 15587171

  • £292.00 / 6 bottles

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Guaranteed endotoxin levels of <0.25EU/mL make it ideal for use in product contact areas
  • GMP manufactured under Grade A laminar flow air flow in a Grade C cleanroom ensures the alcohol, container and packaging are free from contamination and particulates
  • Trigger spray and bag in bottle protected system prevents bottle contents from becoming contaminated during use
  • Bottles can be completely emptied eliminating wastage so no need to discard unused product during shelf life
  • Trigger spray can be set to jet or spray - large droplet size reduces the risk of inhalation and provides good surface coverage
  • Double/triple bagged packed in linear tear packaging so each bag is easy to open even when wearing gloves
  • Sterile via gamma irradiation so suitable for use in Grade A and B cleanrooms
Specifications

Specifications

Ethyl Alcohol
Not Applicable
Colorless
Clear
70% (v/v) Ethanol in 30% WFI
Limited-use
Yes
6 x 1 L
Liquid Spray
Sterile
Not Autoclavable
Denatured ethanol and WFI ( Water for Injection)
Trigger Spray Bottle
Filtered
Sterilizes surfaces in cleanrooms
1 L
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P370+P378-In case of fire: Use dry chemical/carbondioxide to extinguish.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P411-Store at temperatures not exceeding 40°C/104°F.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ETHANOL, PROPAN-2-OL

SDS
Documents

Documents

Certificates
Special Offers

Special Offers