missing translation for 'onlineSavingsMsg'
Learn More

Contec™ Sterile Denatured Ethanol, 70%
BIOPHARMA

Blend of 70% v/v denatured ethanol with 30% purified water. Contec™ Sterile Low Endotoxin Alcohol, 70% is 0.2 micron filtered.

£187.00 - £292.00

Specifications

pH Not Applicable
Sterility Sterile
View More Specs

Products 3
Product Code Brand Container Type Quantity Concentration Price Quantity & Availability  
Product Code Brand Container Type Quantity Concentration Price Quantity & Availability  
16625544
BIOPHARMA
View Documents Promotion Details
Contec™
SBT0570DEW
Trigger Spray Bottle 8 x 0.5 L
£187.00
Pack of 8
Please to purchase this item. Need a web account? Register with us today!
15587171
BIOPHARMA
View Documents Promotion Details
Contec™
Trigger Spray Bottle 6 x 1 L 70% (v/v) Ethanol in 30% WFI
£292.00
6 bottles
Please to purchase this item. Need a web account? Register with us today!
15527171
BIOPHARMA
View Documents Promotion Details
Contec™
Capped Bottle 2 x 5 L 70% (v/v) Ethanol in 30% Purified Water
£257.00
2 bottles
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Guaranteed endotoxin levels of <0.25EU/mL make it ideal for use in product contact areas
  • GMP manufactured under Grade A laminar flow air flow in a Grade C cleanroom ensures the alcohol, container and packaging are free from contamination and particulates
  • Trigger spray and bag in bottle protected system prevents bottle contents from becoming contaminated during use
  • Bottles can be completely emptied eliminating wastage so no need to discard unused product during shelf life
  • Trigger spray can be set to jet or spray - large droplet size reduces the risk of inhalation and provides good surface coverage
  • Double/triple bagged packed in linear tear packaging so each bag is easy to open even when wearing gloves
  • Sterile via gamma irradiation so suitable for use in Grade A and B cleanrooms
Specifications

Specifications

Not Applicable
Sterile
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • CONTEC STERILE DENATURED ETHANOL WITH WFI

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P370+P378-In case of fire: Use dry chemical/coarbindioxide to extinguish.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P411-Store at temperatures not exceeding 40°C/°F.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol, propan-2-ol

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.