Learn More
DWK Life Sciences WHEATON™ CompletePAK™ Sterile Components Seals
The WHEATON™ CompletePAK™ product line is a range of crimp top vials, rubber stoppers and seals that are sterilized ready-to-use and are off- the-shelf, and can be customized for any application reducing your supply chain due to our one source solution capability.
Brand: DWK Life Sciences W986012-S
Description
- Sterile
- Flip-off Seal
Certifications
USP <788>, <85> and <71>; Container Closure Integrity (CCI) certified

Specifications
Blue | |
Sterile | |
Wheaton CompletePAK | |
USP 788, 85, and 71, Container Closure Integrity (CCI) certified | |
5 or 10 mL Serum Vial |
Aluminum | |
220 | |
Sterile Flip Seal | |
20 mm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.