missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Microspec™ Sterile Plastic Inoculation Loops (Volume: 1μL)
Designed for quantitative procedures such as sampling, serial dilutions, as well as for bacterial inoculation.
Brand: Microspec™
Informations supplémentaires : Weight : 18.95000kg
Description
- High Impact Polystyrene construction
- Full length to avoid contamination
- Disposable
- Sterile, aseptically manufactured
- Medium rigidity
- Culture loops (Length: 200mm)
- Smooth edges to avoid tearing and digging in agar
- Volume: 1μL
- Color: Blue
Spécification
Yes | |
High-impact Polystyrene | |
Gamma-irradiated | |
200 mm | |
Blue | |
7.87 in. | |
800 Pouches of 20 | |
Yes |
1 mm | |
7.87 in. | |
0.04 in. | |
1 μL | |
Plastic Culture Loops | |
200 mm | |
Microseal Pouch, Ecopack |