missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Microspec™ Sterile Plastic Inoculation Loops (Volume: 5μL)
Constructed of flexible polystyrene for optimum retrieval of an inoculum from a culture
Brand: Microspec™
Additional Details : Weight : 18.95000kg
Description
- High Impact Polystyrene construction
- Full length to avoid contamination
- Disposable
- Sterile, aseptically manufactured
- Medium rigidity
- Culture loops (Length: 200mm)
- Smooth edges to avoid tearing and digging in agar
- Volume: 5μL
- Color: Blue
Specifications
Yes | |
High-impact Polystyrene | |
Aseptically Manufactured | |
200 mm | |
Plastic Culture Loops | |
200 mm | |
Grip Seal Bag |
2.5 mm | |
7.87 in. | |
0.10 in. | |
Blue | |
7.87 in. | |
50 Pouches of 20 | |
Yes |