missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Socorex™ Sterile Multichannel Reagent Reservoir
Reduce spillage when pouring reagents out of the Socorex™ Sterile Multichannel Reagent Reservoir with pour-off spouts on all four corners. Its trough-within-a-trough design maximizes the amount of liquid accessible to pipet tips when using small amounts of reagent. It is available in 8-channel and 12-channel configurations.Made of polystyrene, it features a trough design and graduation marks on interior walls, .
£110.00 - £138.00
Specifications
Capacity (Metric) | 48 mL |
---|---|
Material | Polystyrene |
Sterility | Sterile |
Type | Reagent Reservoir |
Packaging | Individually wrapped per sterilization |
Product Code | Brand | Channels | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Channels | Price | Quantity & Availability | |||||
10409762
|
Socorex™
330.12.9 |
12 |
£110.00
Pack of 10 |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||||
10060401
|
Socorex™
330.08.9 |
8 |
£138.00
Pack of 10 |
Estimated Shipment: 13-08-2024
Log in to see stock. |
|||||
Description
- U (rounded) bottom
- Single wrapped
- 48mL
- Extra-wide bench for rigidity and stability
- Lot number facilitates traceability.
Specifications
48 mL | |
Sterile | |
Individually wrapped per sterilization |
Polystyrene | |
Reagent Reservoir | |
10 Pack |