missing translation for 'onlineSavingsMsg'
Learn More

Socorex™ Sterile Multichannel Reagent Reservoir

Reduce spillage when pouring reagents out of the Socorex™ Sterile Multichannel Reagent Reservoir with pour-off spouts on all four corners. Its trough-within-a-trough design maximizes the amount of liquid accessible to pipet tips when using small amounts of reagent. It is available in 8-channel and 12-channel configurations.Made of polystyrene, it features a trough design and graduation marks on interior walls, .

Brand:  Socorex™ 330.12.9

 View more versions of this product

Product Code. 10409762

  • £118.00 / Pack of 10
Estimated Shipment: 14-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • U (rounded) bottom
  • Single wrapped
  • 48mL
  • Extra-wide bench for rigidity and stability
  • Lot number facilitates traceability.
Specifications

Specifications

48 mL
Sterile
Individually wrapped per sterilization
12
Polystyrene
Reagent Reservoir
10 Pack
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Socorex™ Sterile Multichannel Reagent Reservoir > Channels: 12

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.