missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Sterile Weigh Boat
GREENER_CHOICE

This sterile weighing boat is flexible and features smooth surfaces to provide accurate pour-outs with minimal sample loss. It is ideal for application in life science research, food, bio-pharmaceutical, environmental, and molecular research environments.

Brand:  Fisherbrand™ 120882

 View more versions of this product

Product Code. 17774824

  • £62.45 / Pack of 200
Estimated Shipment: 12-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Electron beam sterilized to reduce the risk of cross-contamination and compromised samples
  • Made from 100% virgin anti-static polystyrene that is waterproof and reduces the chance of materials sticking to the units
  • Disposable
  • Can be used with microwave moisture analyzers
  • Rounded corners simplify transfer and allow flexing for controlled pouring of liquid, powdered or granular samples
  • Boats are shallow with a wide, flat bottom to resist tipping
  • Color: White
  • FDA/food-grade compliant
Specifications

Specifications

FDA and Sterility
Flat
White
Polystyrene
Sterile
Rounded
Balance
Medium
63.50 x 50.80 x 17.78 mm
50.80 mm
Not Autoclavable
46 mL
Single-use
Hexagonal
Weighing Boat
Weighing
200 Pk.
-10°C to +70°C
63.50 mm
17.78 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers