Learn More
Thermo Scientific™ Sterilin™ Cuvets
Experience top optical quality in spectrophotometry with easy-to-use, polystyrene or acrylic Thermo Scientific™ Sterilin™ Cuvets.
Brand:  Thermo Scientific™ 221S
 View more versions of this product
Product Code. 12751548
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Experience top optical quality in spectrophotometry with easy-to-use, polystyrene or acrylic Thermo Scientific™ Sterilin™ Cuvets.
  • Choice of material: optical quality polystyrene for use at visible wavelengths (340 to 900nm) or optical quality acrylic (PMMA) for use at ultraviolet wavelengths (300 to 900nm)
  • Each case contains cuvets from the same mold cavity to ensure minimal dimensional variation
  • Standard absorption variation between cuvets is better than ±0.005 absorbency units
  • Two sizes available—semi-micro cuvet and macro cuvet
  • Standard pathlength of 10mm
  • Frosted sides on the cuvet provide an ideal labeling and handling area
  • Recessed windows reduce the risk of scratching during use
  • For ease of use an arrow feature indicates the direction of light transmission
  • Packed in expanded polystyrene (EPS) foam trays for optimum protection ensuring scratchfree product at the time of use
Ordering Information
  • Not available in all geographic regions
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Non-sterile
Semi-micro cuvet
Polystyrene
10 mm
22 mm
4.5 mm
Semi-micro Cuvette
1.5 to 3.0 mL
100 pk/ 500 cs
340 to 900 nm
Spectrophotometers and Photometers
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Note: Cuvets should not be used for long-term storage of samples. When using Hydrochloric acid, as the instrument can come under attack from the acid fumes, it is recommended that you use sealing film on the cuvet.