missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sterilin™ Polystyrene Containers, 60mL to 250mL
GREENER_CHOICE

Contain samples in Thermo Scientific™ Sterilin™ Polystyrene Containers with 60mL to 250mL capacities. Available with either plastic or metal flow seal caps.

Brand:  Thermo Scientific™ 190A

64.30 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 11329133

  • £224.00 / Pack of 50

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers

Description

Description

Contain samples in Thermo Scientific™ Sterilin™ Polystyrene Containers with 60mL to 250mL capacities. Available with either plastic or metal flow seal caps.

NOT AVAILABLE IN AMERICA

Features:
  • Aseptically manufactured
  • Leak-tested in accordance with BS EN 14254 Annexe D
  • 125AM, 125BM and 125CM product certified 95kPa-compliant ensuring suitability for sample transport

Note:

For in vitro use only.

Ordering Information:

Product is not available in North America.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Container
Polystyrene
Leak-tested in accordance with BS EN ISO 6717
65.5 mm
No Label
Yes
250 mL
Metal
119.5 mm
50/Cs.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.