missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Sterilization Indicating Labels

Brady™ Sterilization Indicating Labels are steam sterilization indicating polypropylene labels with preprinted headers that displays STERILIZED when exposed to autoclave conditions.

£187.00 - £225.00

Specifications

Adhesive Type Acrylic
Color White
Finish Matte
Material Polypropylene
Print Technology Thermal Transfer
View More Specs

Products 2
Product Code Brand Height (English) Price Quantity & Availability  
Product Code Brand Height (English) Price Quantity & Availability  
17110191
View Documents
Brady™
149273
2 in.
£225.00
Pack of 2000
In Stock
to see stock.
Edge
Add to basket
 
17100191
View Documents
Brady™
149272
1.5 in.
£187.00
Pack of 2000
In Stock
to see stock.
Add to basket
 
Description

Description

  • Ideal for use in sample tracking, inventory management, general tracking, and identification of lab materials that require sterilization
  • Economical solution for identifying vials, tubes, racks and flasks
  • Adheres to stainless steel, glass and polypropylene
  • Withstands temperatures from -320.8 to 249.8° F (-196 to 121°C )

Compliance

RoHS directive 2011/65/EU

Specifications

Specifications

Acrylic
Matte
Thermal Transfer
1 in.
White
Polypropylene
2000/Pk.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Brady™ Sterilization Indicating Labels

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.