missing translation for 'onlineSavingsMsg'
Learn More

Merck Reinforced Membrane Filters

Contain no asbestos or fiberglass; made from non-shedding materials.

Brand:  Merck RW0309000

Code : 71

Additional Details : Weight : 0.15000kg Transport : UN number : 3270 Chem class : 4.1 Pack group : II


 View more versions of this product

Product Code. 10052233

  • £172.34 / Pack of 100
Estimated Shipment: 12-08-2024
to see stock.

Description

Description

Reinforced prefilter membranes can be used to remove contaminants from a variety of liquids and gases. The filters have a high dirt-holding capacity and a low pressure drop, which makes them ideally suited for reducing contaminant levels in advance of sterilizing-grade filters. Unlike traditional prefilter materials that contain asbestos or fiberglass, our reinforced prefilter membranes are made from non-shedding materials. These filters retain contaminants on the surface of the cellulose membrane.

Features & Benefits:
  • Prefilter with high dirt-loading capacity for greater throughput
  • Non-shedding substrate will not unload fibers downstream

Applications: Prefiltration Ahead of Sterilizing-Grade Filters

Specifications

Specifications

Autoclavable
Screen Filter
0.45 μm
Sterile
Submicron clarification, prefiltration for 0.2 to 0.45 μm filters
Hydrophilic
158°F
0.007 in.
Filter Membranes
3.5 in.
9 cm
White
Polymers of Cellulose Ester
0.5 μm
50 mL/min/ cm2
RW03
Plain
70°C
200 μm
Hydrophilic
9 cm
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • X100 Prefilter RW03 90mm

Signal Word
  • Danger

Hazard Category
  • Flammable solid Category 1

Hazard Statement
  • H228-Flammable solid.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P370+P378-In case of fire: Use powder to extinguish.

Supplemental information
  • MIXTURE LIST-Contains : nitrocellulose

SDS
Documents

Documents

Certificates
Special Offers

Special Offers