Learn More
Merck Reinforced Membrane Filters
Contain no asbestos or fiberglass; made from non-shedding materials.
Brand: Merck RW0304700
Code : 71
Additional Details : Weight : 0.01000kg Transport : UN number : 3270 Chem class : 4.1 Pack group : II
Description
Reinforced prefilter membranes can be used to remove contaminants from a variety of liquids and gases. The filters have a high dirt-holding capacity and a low pressure drop, which makes them ideally suited for reducing contaminant levels in advance of sterilizing-grade filters. Unlike traditional prefilter materials that contain asbestos or fiberglass, our reinforced prefilter membranes are made from non-shedding materials. These filters retain contaminants on the surface of the cellulose membrane.Features & Benefits:
- Prefilter with high dirt-loading capacity for greater throughput
- Non-shedding substrate will not unload fibers downstream
Applications: Prefiltration Ahead of Sterilizing-Grade Filters
Specifications
Autoclavable | |
Screen Filter | |
0.45 μm | |
Sterile | |
Submicron clarification, prefiltration for 0.2 to 0.45 μm filters | |
Hydrophilic | |
158°F | |
0.007 in. | |
Filter Membranes | |
1.8 in. | |
4.7 cm |
White | |
Polymers of Cellulose Ester | |
0.5 μm | |
50 mL/min/ cm2 | |
RW03 | |
Plain | |
70°C | |
200 μm | |
Hydrophilic | |
4.7 cm |
Safety and Handling
- Prefltr rw03 47mm 100/pk
Signal Word
- Danger
Hazard Category
- Flammable solid Category 1
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P370+P378-In case of fire: Use powder to extinguish.
Supplemental information
- MIXTURE LIST-Contains : nitrocellulose