Learn More
Sampling Systems™ SteriWare™ Capsule Counting Trays
BIOPHARMA
Reduce the risk of contamination with SteriWare™ Capsule Counting Trays. Ideal for use in hospitals, manufacturing, and pharmaceutical, these tablet counting trays do not require cleaning.
Brand:  Sampling Systems™ 8444A-1S
 View more versions of this product
Product Code. 17574676
  • £95.60 / Pack of 20
Estimated Shipment: 11-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Our capsule counting trays makes accurate sorting and counting easy.
  • They are made from robust, shatterproof PS.
  • They are suitable for use with all capsules from Size 2 to Size 00.
  • Use straight from the heat sealed PE bag.
  • They are manufactured and packed under cleanroom conditions.
  • Made from white food grade PS.
  • Conforms to the latest FDA and EU requirements.
  • BSE/TSE free.
  • Either sterile or non sterile.
Specifications
Specifications
12 mm
Polypropylene
Sterile
260 mm
130 mm
Bag