Learn More
Sampling Systems™ SteriWare™ Disposable Powder Bottle
BIOPHARMA
Designed to meet the requirements of the Pharmaceutical and Hygienic industries. Sampling Systems™ SteriWare™ Disposable Powder Bottle is a high quality, wide mouth bottle that includes a screw top.
Brand:  Sampling Systems™ 8890P-500
 View more versions of this product
Product Code. 15839571
  • £56.00 / Pack of 37
Estimated Shipment: 07-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Moulded and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming material
  • BSE/TSE free
  • Available in 2 sizes
  • Powder tight seal
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • No cross contaminaton
  • No cleaning required
Specifications
Specifications
Bottle
Individually Bagged
Non-sterile
108 mm
Polypropylene
500 mL
106 mm