Learn More
Stoddard Solvent, Thermo Scientific Chemicals
£65.00 - £228.00
Chemical Identifiers
CAS | 8052-41-3 |
---|---|
Molecular Formula | C8H15BrO2 |
Molecular Weight (g/mol) | 223.11 |
MDL Number | MFCD00132767 |
InChI Key | IMSPZLCTPSXORJ-UHFFFAOYSA-N |
Synonym | 6-bromo-1-hexanol acetate, 1-acetoxy-6-bromohexane, stoddard solvent, 1-hexanol,6-bromo-,acetate, neo-clear r xylene substitute, w/w benzene, acetic acid 6-bromohexyl ester, 6-acetoxy-1-bromohexane |
PubChem CID | 11806291 |
IUPAC Name | 6-bromohexyl acetate |
SMILES | CC(=O)OCCCCCCBr |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10474701
|
Thermo Scientific Acros
419730010 |
1 kg |
£65.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15284734
|
Thermo Scientific Acros
419730050 |
5 kg |
£228.00
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
8052-41-3 | |
223.11 | |
IMSPZLCTPSXORJ-UHFFFAOYSA-N | |
11806291 | |
CC(=O)OCCCCCCBr |
C8H15BrO2 | |
MFCD00132767 | |
6-bromo-1-hexanol acetate, 1-acetoxy-6-bromohexane, stoddard solvent, 1-hexanol,6-bromo-,acetate, neo-clear r xylene substitute, w/w benzene, acetic acid 6-bromohexyl ester, 6-acetoxy-1-bromohexane | |
6-bromohexyl acetate |
Specifications
-40.0°C | |
0.7870g/mL | |
>38°C | |
Solvent | |
C8H15BrO2 | |
1268 | |
Solubility in water: insoluble. | |
CC(=O)OCCCCCCBr | |
223.11 | |
Stoddard solvent |
8052-41-3 | |
149.0°C to 213.0°C | |
Glass Bottle | |
0.787 | |
MFCD00132767 | |
6-bromo-1-hexanol acetate, 1-acetoxy-6-bromohexane, stoddard solvent, 1-hexanol,6-bromo-,acetate, neo-clear r xylene substitute, w/w benzene, acetic acid 6-bromohexyl ester, 6-acetoxy-1-bromohexane | |
IMSPZLCTPSXORJ-UHFFFAOYSA-N | |
6-bromohexyl acetate | |
11806291 |
Safety and Handling
GHS P Statement May be fatal if swallowed and enters airways. Toxic to aquatic life with long lasting effects. Flammable liquid and vapor. Repeated exposure may cause skin dryness or cracking.
GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Do not breathe dust/fume/gas/mist/vapors/spray. Do not get in eyes, on skin, or on clothing. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Do NOT induce vomiting.
Danger
missing translation for 'einecsNumber' : 232-489-3
missing translation for 'rtecsNumber' : WJ8925000
missing translation for 'tsca' : TSCA
- Stoddard solvent
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Flammable liquid Category 3
- Specific target organ toxicity Category 1
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
- H372-Causes damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- H226-Flammable liquid and vapour.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P262-Do not get in eyes, on skin, or on clothing.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
RUO – Research Use Only