Learn More
Stopcock Assembly and Cover for Nalgene™ Transparent Polycarbonate Classic Design Desiccator
GREENER_CHOICE
Get replacement parts for the Nalgene™ Transparent Polycarbonate Classic Design Desiccator
£371.00
Products 1
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
10326231
GREENER_CHOICE
View Documents
Thermo Scientific Nalgene
715311-0250
Cover for port on dessicator, polycarbonate
£371.00
Pack of 2
Estimated Shipment: 23-06-2025
to see stock.
Add to basket
Description
Description
Get the Stopcock Assembly and Cover replacement parts for the Nalgene™ Transparent Polycarbonate Classic Design Desiccator, Cat #5311-0250. The Nalgene™ Transparent Polycarbonate Classic Design Desiccator maintains an airtight seal without the need of vacuum grease and can be used under full-vacuum and non-vacuum conditions, and is an excellent alternative to glass.
Description:
  • Replacement stopcock for Nalgene desiccator item number 5311-0250 
  • Replacement cover for Nalgene desiccator item number 5311-0250 
Please recycle the corrugated cartons and LDPE bags in which your Nalgene products are packaged.