missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gosselin™ Straight Specimen Containers with Green Cap
CORNING GOSSELIN™ Straight Container, Specimen Vials, PP come in different sizes and quantities, and are available sterile and non-sterile.
Brand: Gosselin™ TP52C-014
This item is not returnable.
View return policy
Beschreibung
- Clear and break-resistant polypropylene (PP)
- Available in different sizes and quantities
- Color-coded screw caps
Spezifikation
Specimen Container | |
Polypropylene | |
Non-sterile | |
Green | |
Clear | |
380/Cs. |
Yes | |
Straight | |
125 mL | |
Screw Cap | |
No Label | |
380/Pk. |