missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences Strep-Tactin™ 4Flow™ high capacity 50% suspension

Strep-Tactin™ 4Flow™ high capacity 50% Suspension to purify Strep-tag™II or Twin-Strep-tag™ fusion proteins

Brand:  IBA Lifesciences 2-1250-500

 View more versions of this product

Product Code. 30117606

  • £19220.00 / 1000mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Strep-Tactin™ 4Flow™ high capacity is a resin for purification of strep-tagged proteins via affinity chromatography
  • Product is optimized for column-based use and suitable for HPLC/FPLC
  • Recommended for all protein classes due to the broad pH compatibility, excellent pressure stability, outstanding target purity (>95%), and high binding affinity
  • 4% crosslinked agarose beads coupled to Strep-Tactin™ for straightforward purification of proteins with Strep-tag™II or a Twin-Strep-tag™
  • Targeted elution: no tedious titration steps required and multiple regeneration cycles possible
  • Ideal alternative for proteins that are difficult to elute from Strep-Tactin™XT 4Flow™ resins
  • Biotinylated proteins also bind to Strep-Tactin™ 4Flow™ high capacity and need to be blocked using BioLock
Specifications

Specifications

20 mg/mL resin
50% suspension
50 to 150 μm
1000 mL
Proteins tagged with Twin-Strep-tag™; Strep-tag™II; cross reactivity: biotin
Affinity chromatography equipment
Agarose
Bottle
1000 mL
4% agarose, crosslinked
Resin Slurry
Tetrameric protein
Bulk Resin
Twin-Strep-tag™; Strep-tag™II; cross reactivity: biotin
Resin sufficient for 500 times 1 mL gravity flow column
Suspension
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers