missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences Strep-Tactin™ Sepharose™ Resin

Strep-Tactin™ Sepharose™ resin is a 50% suspension to purify Strep-tag™II or Twin-Strep-tag™ fusion proteins

Brand:  IBA Lifesciences 2-1201-010

 View more versions of this product

Product Code. 12846436

  • £722.00 / 20mL
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Strep-Tactin™ Sepharose™ is a 4% agarose coupled with the streptavidin variant Strep-Tactin™ and applicable for purification of Strep-tag™II or Twin-Strep-tag™ fusion proteins.
  • Due to the low concentrated agarose, it is particularly suitable for large proteins, especially if the affinity to Strep-Tactin™XT 4Flow™ is too high.
  • The wide-meshed polymer allows for the simple attachment of larger proteins to the stationary phase.
  • The suspension can be used for packing of gravity flow columns since the resin is optimized for column affinity chromatography, rather than for batch purification.
  • To pack a column with 1 ml resin bed volume, 2 ml 50% suspension is necessary.

Please note that Strep-Tactin™ Sepharose™ is not compatible with HPLC/FPLC due to the low pressure stability.

Specifications

Specifications

Strep-Tactin Sepharose Suspension (50%)
4°C
Strep-Tactin Sepharose
20 mL
Preferentially for Gravity Flow Purification of Strep-Tag Fusion Proteins
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.