Learn More
IBA Lifesciences Strep-Tactin™XT 4Flow™ Resin
Strep-Tactin™XT 4Flow™ resin is a 50% suspension to purify Strep-tag™II or Twin-Strep-tag™ fusion proteins
Brand: IBA Lifesciences 2-5010-002
This item is not returnable.
View return policy
Description
- Strep-Tactin™XT 4Flow™ is a 4% agarose coupled with the streptavidin variant Strep-Tactin™XT and applicable for purification of Strep-tag™II or Twin-Strep-tag™ fusion proteins.
- Due to the broad pH compatibility, excellent pressure stability, outstanding target purity (> 95%), and highest binding affinity, the resin is recommended for all protein classes, especially for low abundant or challenging proteins.
- The low concentrated agarose allows for the simple attachment of larger proteins to the stationary phase, making it particularly suitable for purification of high-molecular-weight targets.
- In addition, Strep-Tactin™XT 4Flow™ enables the application of denaturing agents such as urea.
- The suspension can be used for packing of gravity flow columns since the resin is optimized for column affinity chromatography, rather than for batch purification.
- To pack a column with 1 ml resin bed volume, 2 ml 50% suspension are necessary.
Specifications
4% agarose, crosslinked | |
4 mL | |
Pre-packed Gravity Flow Column |
50 to 150 μm | |
Twin-Strep-Tag™ and Strep-tag™II |