Learn More
IBA Lifesciences™ Strep-Tactin™ XT BLI Coupling Kit for Coating of Biosensors
The XT BLI Coupling Kit can be used for coating BLI biosensors for reversible capture and analysis of proteins.
Brand:  IBA Lifesciences™ 2-4380-000
Product Code. 17580655
  • £709.67 / 1 set
Explore more special offers
Description
Description
Strep-Tactin™ XT BLI Coupling Kit is intended for site-directed, reversible capture of Twin-Strep-tag™ fusion proteins for biomolecular interaction analysis using Bio-Layer Interferometry (BLI). The pM affinity of Strep-Tactin™ XT for Twin-Strep-tag™ fusion proteins allows specific capture even from complex samples, such as culture supernatants. Coated biosensors can be easily regenerated by application of the provided Regeneration Buffer without loss of the Strep-Tactin™ XT coating, which enables multiple uses.
  • Provides Strep-Tactin™ XT, Immobilization Buffer, Ethanolamine, EDC, and s-NHS sufficient for coating of 96 biosensors (1 tray)
Specifications
Specifications
In Low pM Range
Bio-Layer Interferometry (BLI) Systems
1 Kit
Strep-Tactin™ XT
Optical Biosensing Technology
Yes
Biomolecular Interaction Analysis
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Show More
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
IBA Lifesciences™ Strep-Tactin™ XT BLI Coupling Kit for Coating of Biosensors > XT BLI Coupling Kit; Binding Strength: In Low pM Range; Detection Method: Optical Biosensing Technology; Ligand Type: Strep-Tactin™ XT; For Use With (Equipment): Bio-Layer Interferometry (BLI) Systems