missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences Strep-Tactin™XT Conjugate
GREENER_CHOICE

Product Code. p-7230581 Shop All IBA Lifesciences Products
Click to view available options
Label or Dye:
DY-488
DY-549
DY-649
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 16401475

Brand: IBA Lifesciences 21562050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Strep-Tactin™XT conjugates with fluorescent label to detect Strep-tag™II and Twin-Strep-tag™ fusion proteins

Strep-Tactin™XT is a streptavidin variant, which binds to the Strep-tag™II and Twin-Strep-tag™ with high affinity and specificity.

It is available as conjugate with fluorescent dyes (DY-488, DY-549, DY-649) for fast and directed detection of N-terminally, C-terminally as well as internally tagged Strep-tag™II or Twin-Strep-tag™ fusion proteins in applications like ELISA, FACS, immunohistochemistry, and immunocytochemistry.

The absorption/emission maxima of the fluorescent dyes are:
• DY-488: 493/518 nm
• DY-549: 550-560/575 nm
• DY-649: 655/676 nm

Especificaciones

Capacity (Metric) 0.10 mL
Concentration 0.5 mg/mL
Formulation Strep-Tactin™XT conjugated with DY-488
Kit Contents Solution in PBS; 0.01 % NaN3; 1 mg/ mL BSA
Content And Storage Undiluted: 2°C to 8°C, protect from light; Diluted: -25°C to -15°C
Quantity 50 μg
For Use With (Application) ELISA, FACS, western blot, immunofluorescence
Target Strep-tag™II and Twin-Strep-tag™
Type Conjugate
Label or Dye DY-488

For Research Use Only.

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado