missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences Strep-Tactin™XT Superflow™ Cartridge

Strep-Tactin™XT Superflow™ cartridge is a HPLC/FPLC cartridge to purify Strep-tag™II or Twin-Strep-tag™ fusion proteins

£88.97 - £230.58

Specifications

Binding Capacity 5.7 mg protein/ mL resin
Quantity 1 Cartridge
For Use With (Application) Protein Purification
Sufficient For Chromatpgraphy workstations with 10 to 32 connections
View More Specs

Products 2
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
15583077
View Documents
IBA Lifesciences
2-4021-001
1 x 1 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15728207
View Documents
IBA Lifesciences
2-4022-001
1 x 5 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Strep-Tactin™XT Superflow™ cartridges contain a 6% agarose coupled with the streptavidin variant Strep-Tactin™XT and are applicable for purification of Strep-tag™II or Twin-Strep-tag™ fusion proteins with HPLC/FPLC devices like Äkta™ systems (Cytiva).
  • In comparison to Strep-Tactin™ Superflow™, Strep-Tactin™XT Superflow™ has a higher affinity for Strep-tag™ fusion proteins, which leads to an improved immobilization and purification.
  • In addition, Strep-Tactin™XT Superflow™ allows the application of denaturing agents such as urea during the purification procedure.
  • As with any other Strep-Tactin™XT resins, the unspecific protein binding is extremely low, which results in an outstanding purity of the target protein. 
Specifications

Specifications

5.7 mg protein/ mL resin
1 Cartridge
Protein Purification
Chromatpgraphy workstations with 10 to 32 connections
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.