missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences StrepMAB-Classic Conjugate
GREENER_CHOICE

StrepMAB-Classic conjugates are monoclonal antibodies with different conjugates to detect Strep-tag™II and Twin-Strep-tag™ fusion proteins

Brand:  IBA Lifesciences 2-1563-050

 View more versions of this product

Product Code. 16411475

  • £288.00 / 50µg
Estimated Shipment: 14-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

StrepMAB-Classic is a highly selective, murine monoclonal antibody for detection of Strep-tag™II and Twin-Strep-tag™ fusion proteins in applications like western blots, ELISA, FACS, immunohistochemistry, and immunocytochemistry.

It is available coupled to fluorescent dyes (DY-488, DY-549, DY-649). 

The absorption/emission maxima of the fluorescent dyes are:
• DY-488: 493/518 nm
• DY-549: 550-560/575 nm
• DY-649: 655/676 nm

Please note: For capturing of Strep-tag™II and Twin-Strep-tag™ fusion proteins on solid phases, the use of StrepMAB-Immo or Strep-Tactin™XT is recommended due to the binding affinity in the pM range.

Specifications

Specifications

0.10 mL
Solution in PBS; 0.01 % NaN3; 1 mg/ mL BSA
50 μg
Strep-tag™II and Twin-Strep-tag™
DY-488
0.5 mg/mL
Undiluted: 2°C to 8°C, protect from light; Diluted: -25°C to -15°C
ELISA, western blot, immunofluorescence
Conjugate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.