missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences StrepMAB-Classic Conjugate
GREENER_CHOICE

StrepMAB-Classic conjugates are monoclonal antibodies with different conjugates to detect Strep-tag™II and Twin-Strep-tag™ fusion proteins

Brand:  IBA Lifesciences 2-1566-050

 View more versions of this product

Product Code. 16441475

  • £267.00 / 50µg
Estimated Shipment: 11-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

StrepMAB-Classic is a highly selective, murine monoclonal antibody for detection of Strep-tag™II and Twin-Strep-tag™ fusion proteins in applications like western blots, ELISA, FACS, immunohistochemistry, and immunocytochemistry.

It is available coupled to fluorescent dyes (DY-488, DY-549, DY-649). 

The absorption/emission maxima of the fluorescent dyes are:
• DY-488: 493/518 nm
• DY-549: 550-560/575 nm
• DY-649: 655/676 nm

Please note: For capturing of Strep-tag™II and Twin-Strep-tag™ fusion proteins on solid phases, the use of StrepMAB-Immo or Strep-Tactin™XT is recommended due to the binding affinity in the pM range.

Specifications

Specifications

0.10 mL
Solution in PBS; 0.01 % NaN3; 1 mg/ mL BSA
50 μg
Strep-tag™II and Twin-Strep-tag™
DY-549
0.5 mg/mL
Undiluted: 2°C to 8°C, protect from light; Diluted: -25°C to -15°C
ELISA, western blot, immunofluorescence
Conjugate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.