missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences StrepMAB-Immo Conjugate
GREENER_CHOICE

Product Code. p-7230576 Shop All IBA Lifesciences Products
Click to view available options
Label or Dye:
DY-488
DY-549
DY-649
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 16421475

Brand: IBA Lifesciences 21564050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

StrepMAB-Immo conjugates are monoclonal antibodies with different fluorescent conjugates to detect Strep-tag™II and Twin-Strep-tag™ fusion proteins

StrepMAB-Immo fluorescenct conjugates are murine, high-affinity monoclonal antibodies, which are especially suited for detection of Strep-tag™II and Twin-Strep-tag™ fusion proteins in applications like ELISA, FACS, immunohistochemistry, and immunocytochemistry.

The nearly irreversible binding is validated for fusion proteins carrying a C- or N-terminal Strep-tag™II providing an N-terminal extension by a SerAla linker (recombinant protein-SA-WSHPQFEK or SA-WSHPQFEK-recombinant protein).

Proteins containing Strep-tag™II with N-terminal sequences other than SerAla are bound with reduced affinity in the cases tested so far, the Twin-Strep-tag™ always contains the SerAla linker (WSHPQFEK-GGGS-GGGS-GGSA-WSHPQFEK). StrepMAB-Immo is available as conjugate with different fluorescent dyes (DY-488, DY-549, DY-649).

The absorption/emission maxima of the fluorescent dyes are:
• DY-488: 493/518 nm
• DY-549: 550-560/575 nm
• DY-649: 655/676 nm

Please note: For detection of Strep-tag™II or Twin-Strep-tag™ fusion proteins in western blots, the use of StrepMAB-Classic or Strep-Tactin™ is recommended.

Specifications

Capacity (Metric) 0.10 mL
Concentration 0.5 mg/mL
Kit Contents Solution in PBS; 0.01 % NaN3; 1 mg/ mL BSA
Content And Storage Undiluted: 2°C to 8°C, protect from light; Diluted: -25°C to -15°C
Quantity 50 μg
For Use With (Application) ELISA, FACS, immunofluorescence
Target Strep-tag™II and Twin-Strep-tag™
Type Conjugate
Label or Dye DY-488

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.