missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ Streptavidin Acrylamide
streptavidin acrylamide
Brand: Molecular Probes™ S21379
Code : 50
Additional Details : Weight : 0.00525kg
Description
Streptavidin acrylamide can be co-polymerized with acrylamide on polymeric surfaces to create a uniform monolayer of the immobilized streptavidin. The streptavidin can then bind and isolate biotinylated-ligands, including probes, enzymes, antibodies and drugs.Specifications
Store in freezer -5°C to -30°C. | |
Liquid Suspension | |
Acrylamide | |
Affinity |
Room Temperature | |
Streptavidin Acrylamide | |
Streptavidin |
For Research Use Only. Not for use in diagnostic procedures.