Learn More
Cytiva Streptavidin Mag Sepharose™ Magnetic Bead
For simple and efficient enrichment of target proteins by immunoprecipitation and purification of biotinylated biomolecules
Brand: Cytiva 28-9857-99
326.32 GBP valid until 2024-12-31
Use promo code "21979" to get your promotional price.
View more versions of this product
Description
Streptavidin Mag Sepharose uses the strong interaction between biotin and streptavidin ligand, which is immobilized on magnetic beads.
- High-capacity small-scale purification of proteins from cell lysates and biological fluids
- Provides high purity and yield
- Allows easy parallel screening of proteins with high repeatability
- Simple enrichment of proteins from small or large sample volumes (low μl to high ml scale)
- The beads are available in two pack sizes: 2 × 1mL 10% medium slurry and 5 × 1mL 10% medium slurry
- A milliliter of 10% medium slurry is the same as 100 μl sedimented medium which is sufficient for 20 purification runs according to the recommended immunoprecipitation protocol
- Binding capacity: >300μg biotinulated BSA/mL of medium slurry
- Stability: stable
- Immunoprecipitation
- Purification
- Biomarker reseach
Specifications
White | |
4°C to 8°C | |
For simple and efficient enrichment of target Proteins by immunoprecipitation and purification of Biotinylated biomolecules |
5 x 1 mL | |
Liquid |
Safety and Handling
- Streptavidin Mag Sepharose
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-66.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
- UNKNOWN AQUATIC-Contains 50% of components with unknown hazards to the aquatic environment.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.