missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Streptavidin Protein, HRP
Brand: Thermo Scientific™ 21124
397.10 GBP valid until 2024-12-31
Use promo code "21979" to get your promotional price.
This item is not returnable.
View return policy
Description
Streptavidin is biotin-binding protein that was originally isolated from Streptomyces avidinii. In contrast to avidin, streptavidin has no carbohydrate and has a mildly acidic pI of 5. Invitrogen Streptavidin products use a recombinant form of streptavidin having a mass of 53,000 daltons and a near-neutral pI. Streptavidin is a tetrameric protein, with each subunit binding one molecule of biotin with affinity similar to that of avidin. Guanidinium chloride will dissociate avidin and streptavidin into subunits, but streptavidin is more resistant to dissociation.Formulation: 2mg purified protein in PBS and no preservative; pH 6.8
This product has been successfully used in ELISA, Immunohistochemistry and Western Blot procedures.
Specifications
HRP | |
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, but do not store above -20°C. | |
Solid | |
Streptavidin Conjugate | |
HRP (Horseradish Peroxidase) |
2 mg | |
Ambient Temperature | |
Pierce™ | |
Biotin |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction