missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Streptavidin Protein, HRP
£242.00 - £728.00
Specifications
Conjugate | HRP |
---|---|
Content And Storage | Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, but do not store above -20°C. |
Shipping Condition | Ambient Temperature |
Form | Solid |
Product Line | Pierce™ |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
10086023
|
Thermo Scientific™
21126 |
1mg; Label: HRP |
£242.00
1mg |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||||
10698084
|
Thermo Scientific™
21124 |
2mg; Label: HRP |
£418.00
2mg |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
10197673
|
Thermo Scientific™
21127 |
5mg; Label: HRP |
£728.00
5mg |
Estimated Shipment: 10-07-2024
Log in to see stock. |
|||||
Description
Streptavidin is biotin-binding protein that was originally isolated from Streptomyces avidinii. In contrast to avidin, streptavidin has no carbohydrate and has a mildly acidic pI of 5. Invitrogen Streptavidin products use a recombinant form of streptavidin having a mass of 53,000 daltons and a near-neutral pI. Streptavidin is a tetrameric protein, with each subunit binding one molecule of biotin with affinity similar to that of avidin. Guanidinium chloride will dissociate avidin and streptavidin into subunits, but streptavidin is more resistant to dissociation. This product has been successfully used in ELISA, Immunohistochemistry and Western Blot procedures.Specifications
HRP | |
Ambient Temperature | |
Pierce™ | |
Biotin |
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, but do not store above -20°C. | |
Solid | |
Streptavidin Conjugate | |
HRP (Horseradish Peroxidase) |
For Research Use Only. Not for use in diagnostic procedures.