Learn More
Cytiva Streptavidin Sepharose High Performance Resin
Affinity chromatography resin designed for fast and reliable, high resolution purification of biotinylated biomolecules.
Brand: Cytiva 90100484
Description
- Streptavidin immobilized to Sepharose High Performance for purification of biotinylated compounds
- Extremely useful for exploiting either the strong interactions of biotin and streptavidin or the somewhat weaker interactions of 2-iminobiotin and streptavidin.
- 34 μm bead size for improved resolution
Purified Streptavidin isolated from Streptomyces avidinii is immobilised on Sepharose High Performance. The base matrix is a rigid, highly cross-linked beaded agarose with high chemical stability. The immobilised streptavidin binds biotin and biotinylated substances through affinity chromatography.

Specifications
25 mL | |
4 to 9 | |
25 mL |
Highly cross-linked agarose, 6% | |
4 to 8 °C, 20% Ethanol |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- Streptavidin Sepharose High Performance
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.