missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Streptex™ Latex Agglutination Test

Isolate and identify a variety of streptococci with a simple enzyme extraction method that detects Lancefield groups A, B, C, D, F and G.

Brand:  Thermo Scientific™ R30164701

 View more versions of this product

Product Code. 13255639

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

2°C to 8°C
Yes
Streptococcus Test Kit
Test latex, extraction enzyme, polyvalent positive control, disposable reaction cards, disposable mixing sticks
Lancefield A, B, C, D, F
Staphylococcus aureus
Streptex Latex Agglutination Test
Streptococci
Enzyme Immunoassay Detection
Polyvalent
200 Tests/Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6188963 Certificate TypeCertificate of Analysis Date10/02/2025 Catalog NumberR30164701
Lot Number6165448 Certificate TypeCertificate of Analysis Date04/11/2024 Catalog NumberR30164701
Lot Number6136181 Certificate TypeCertificate of Analysis Date20/08/2024 Catalog NumberR30164701
Lot Number3807758 Certificate TypeCertificate of Analysis Date18/06/2024 Catalog NumberR30164701
Lot Number3774341 Certificate TypeCertificate of Analysis Date11/04/2024 Catalog NumberR30164701
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Streptex™ Latex Agglutination Test >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.