missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva StrepTrap™ HP Prepacked Columns
Purify Strep(II)-tagged recombinant proteins by affinity chromatography. Cytiva Life Sciences™ StrepTrap™ HP Prepacked Columns are packed with a robust and stable Sepharose™ based medium which results in high resolution, sharp concentrated peaks, and reproducible purifications. The binding capacity is also high.
Brand: Cytiva 28-9075-46
This item is not returnable.
View return policy
Description
- Prepacked with StrepTactin Sepharose High Performance
- Binding capacity Approx. 6 mg Strep II-tagged protein/ml medium
- Maximum pressure: 3 bar
- Column material: polypropylene
- Compatible with a wide range of reducing agents, detergents, denaturants, and other additives
- Generated with 0.5M NaOH
- 1 and 5mL columns
Media
- Ligand: Streptactin
- Matrix: 6% cross-linked agarose
- Average particle size: 34μm
- pH stability working range: >7
1mL columns
- Bed dimensions: 7 x 25mm
- Flow velocity: <4mL/min.
5mL columns
- Bed dimensions: 16 x 25mm
- Flow velocity: <20mL/min.
Specifications
StrepTactin Sepharose HP prepacked column | |
0.27 in. | |
5 x 1mL | |
25mm | |
Polypropylene | |
3 bar | |
34μm | |
2° to 8°C | |
Purification of Strep (II) -tagged recombinant Proteins by affinity chromato graphy |
1mL | |
7mm | |
0.98 in. | |
<4 mL/min. | |
StrepTactin (∼5mg/mL medium) | |
7 | |
6% Highly Cross-linked Agarose | |
Ion Exchange Column |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva StrepTrap™ HP Prepacked Columns > Flow rate: less than 4mL/min.; Bed Dimensions: 7 x 25mm; 5 x 1mL
Spot an opportunity for improvement?Share a Content Correction