missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva StrepTrap™ HP Prepacked Columns
Purify Strep(II)-tagged recombinant proteins by affinity chromatography. Cytiva Life Sciences™ StrepTrap™ HP Prepacked Columns are packed with a robust and stable Sepharose™ based medium which results in high resolution, sharp concentrated peaks, and reproducible purifications. The binding capacity is also high.
Brand: Cytiva 28-9075-48
This item is not returnable.
View return policy
Description
- Prepacked with StrepTactin Sepharose High Performance
- Binding capacity Approx. 6 mg Strep II-tagged protein/ml medium
- Maximum pressure: 3 bar
- Column material: polypropylene
- Compatible with a wide range of reducing agents, detergents, denaturants, and other additives
- Generated with 0.5M NaOH
- 1 and 5mL columns
Media
- Ligand: Streptactin
- Matrix: 6% cross-linked agarose
- Average particle size: 34μm
- pH stability working range: >7
1mL columns
- Bed dimensions: 7 x 25mm
- Flow velocity: <4mL/min.
5mL columns
- Bed dimensions: 16 x 25mm
- Flow velocity: <20mL/min.
Specifications
StrepTactin Sepharose HP prepacked column | |
0.62 in. | |
5 x 5mL | |
25mm | |
Polypropylene | |
3 bar | |
34μm | |
4° to 8°C | |
Purification of Strep (II) -tagged recombinant Proteins by affinity chromato graphy |
5mL | |
16mm | |
0.98 in. | |
<20 mL/min. | |
StrepTactin (∼5mg/mL medium) | |
7 | |
6% Highly Cross-linked Agarose | |
Ion Exchange Column |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- StrepTrap HP, 5 ml
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : Ethanol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva StrepTrap™ HP Prepacked Columns > Flow rate: less than 20mL/min.; Bed Dimensions: 16 x 25mm; 5 x 5mL
Spot an opportunity for improvement?Share a Content Correction