missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva StrepTrap XT Chromatography Column
StrepTrap XP is a column packed with Strep-Tactin™ XT Sepharose resin to purify Strep-tag™ II and Twin-Strep-tag™ recombinant proteins.
£216.00 - £2239.00
Specifications
Height (Metric) | 25 mm |
---|---|
Column Type | Affinity Column |
For Use With (Application) | Purification of recombinant proteins tagged with Strep-tag™ II and Twin-Strep-tag™ |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16522542
|
Cytiva
29401317 |
1 x 1 mL |
£216.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16456149
|
Cytiva
29401320 |
5 x 1 mL |
£548.03
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16466149
|
Cytiva
29401322 |
1 x 5 mL |
£553.28
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16476149
|
Cytiva
29401323 |
5 x 5 mL |
£2239.00
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- For use with Strep-Tactin™ XT Sepharose™
- Prepacked in chromatography columns for convenience
Specifications
25 mm | |
Affinity Column | |
Purification of recombinant proteins tagged with Strep-tag™ II and Twin-Strep-tag™ |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- StrepTrap XT, 1 x 1 ml
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva StrepTrap XT Chromatography Column