Learn More
Corning™ Stripwell™ Microplates
Designed and treated to provide uniform binding, high optical clarity, and low background interference. Corning™ Stripwell™ Microplates provide a superior solution for in vitro diagnostic assays.
Brand: Corning™ 2481
Code : 67
Informations supplémentaires : Weight : 0.04934kg
Description
Corning™ Stripwell™ microplates may be used in a variety of laboratory assays. The flat bottom strips are designed to easily break apart and are pre-assembled in an “egg-crate” style strip holder that allows each individual well to be positioned back into the microplate once broken.
- Stripwell microplates have 96-well flat bottom polystyrene format.
- Low volume and standard Stripwell microplates have well volumes of 190 μL and 360 μL, respectively.
- 1 x 8 strips are designed to fit only one way into the strip holder, eliminating the chance of misorientation.
Spécification
High Binding | |
Without Lid | |
High Bind | |
Polystyrene | |
75 to 125 μL | |
High Binding | |
Round | |
100/Cs. | |
100/Cs. |
Strip Plate | |
8 x 12 | |
Clear | |
96 | |
Non-sterile | |
190 μL | |
In-vitro diagnostic assays | |
25/Pk. |