Learn More
Corning™ Stripwell™ Microplates
Designed and treated to provide uniform binding, high optical clarity, and low background interference. Corning™ Stripwell™ Microplates provide a superior solution for in vitro diagnostic assays.
Brand: Corning™ 3924
654.48 GBP valid until 2024-12-31
Use promo code "22007" to get your promotional price.
Description
Corning™ Stripwell™ microplates may be used in a variety of laboratory assays. The flat bottom strips are designed to easily break apart and are pre-assembled in an “egg-crate” style strip holder that allows each individual well to be positioned back into the microplate once broken.
- Stripwell microplates have 96-well flat bottom polystyrene format.
- Low volume and standard Stripwell microplates have well volumes of 190 μL and 360 μL, respectively.
- 1 x 8 strips are designed to fit only one way into the strip holder, eliminating the chance of misorientation.
Order Info
Bulk packed. Cat. No. 07-200-24 has individual 1 x 8 strips without frame.
Certifications
Cat. Nos. 07-200-31 and -32 have certified surface chemistry.
Specifications
High Binding | |
Without Lid | |
High Binding | |
Polystyrene | |
75 to 200 μL | |
360 μL | |
In-vitro diagnostic assays | |
25/Pk. |
Strip Plate | |
8 x 12 | |
Black | |
96 | |
Non-sterile | |
Round, Flat Bottom (F-well) | |
100/Cs. | |
100/Cs. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.