missing translation for 'onlineSavingsMsg'
Learn More
Strontium bromide hexahydrate, 95%, Thermo Scientific Chemicals Product Code.: 11324879

Strontium bromide hexahydrate, 95%, Thermo Scientific Chemicals

Product Code. 11324879
100 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 11324879

Brand: Thermo Scientific Alfa Aesar 072105.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Strontium bromide hexahydrate is used in flares and also used in some pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-53-9
Molecular Formula Br2H12O6Sr
Molecular Weight (g/mol) 355.518
MDL Number MFCD00150361
InChI Key FLMJUJXBFKFYOZ-UHFFFAOYSA-L
Synonym unii-7i32n2ud6w, strontium dibromide hexahydrate, strontium bromide, reagent, strontium 2+ hexahydrate dibromide, strontium 2+ ion hexahydrate dibromide
PubChem CID 165644
IUPAC Name strontium;dibromide;hexahydrate
SMILES O.O.O.O.O.O.[Br-].[Br-].[Sr+2]

Specifications

Physical Form Crystalline
Quantity 100 g
Assay Percent Range 95%
Solubility Information Soluble in water,alcohol
Formula Weight 355.53 (247.44 Anhydrous)
Sensitivity Hygroscopic
Density 2.386 g/mL
Chemical Name or Material Strontium bromide hexahydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberT31J006 Certificate TypeCertificate of Analysis Date09/08/2023 Catalog Number072105.36,072105.22
Lot NumberN07I051 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number072105.36,072105.22
Lot NumberA31T037 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number072105.36,072105.22
Lot NumberR31D043 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number072105.36,072105.22
Lot NumberT17E052 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number072105.36,072105.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Strontium bromide hexahydrate, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.