Learn More
Strontium bromide, ultra dry, 99.995% (metals basis), Thermo Scientific Chemicals
£76.10 - £293.00
Chemical Identifiers
CAS | 10476-81-0 |
---|---|
Molecular Formula | Br2Sr |
Molecular Weight (g/mol) | 247.428 |
MDL Number | MFCD00049548 |
InChI Key | YJPVTCSBVRMESK-UHFFFAOYSA-L |
Synonym | acmc-20ajmt, strontium bromide srbr2, strontium 2+ ion dibromide |
PubChem CID | 5360351 |
IUPAC Name | strontium;dibromide |
SMILES | [Br-].[Br-].[Sr+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11358567
|
Thermo Scientific Alfa Aesar
035717.03 |
1 g |
£76.10
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11368567
|
Thermo Scientific Alfa Aesar
035717.06 |
5 g |
£293.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used as sedatives in medicine and analytical reagents, for facile synthesis of poly(phenylcarbyne), which is a precursor to diamond-like carbon.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Chemical Identifiers
10476-81-0 | |
247.428 | |
YJPVTCSBVRMESK-UHFFFAOYSA-L | |
5360351 | |
[Br-].[Br-].[Sr+2] |
Br2Sr | |
MFCD00049548 | |
acmc-20ajmt, strontium bromide srbr2, strontium 2+ ion dibromide | |
strontium;dibromide |
Specifications
643°C | |
White | |
Br2Sr | |
MFCD00049548 | |
acmc-20ajmt, strontium bromide srbr2, strontium 2+ ion dibromide | |
YJPVTCSBVRMESK-UHFFFAOYSA-L | |
strontium;dibromide | |
5360351 | |
99.995% | |
Ampule | |
4.216 g/mL | |
Strontium bromide, Ultra dry |
10476-81-0 | |
Powder | |
SrBr2 | |
14,8837 | |
Soluble in water,carbinol,ethanol,amyl alcohol. Insoluble in ether. | |
[Br-].[Br-].[Sr+2] | |
247.428 | |
247.44 | |
Odorless | |
Hygroscopic | |
(metals basis) |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 233-969-5
missing translation for 'rtecsNumber' : WK8050000
missing translation for 'tsca' : Yes
missing translation for 'storageNote' :
- Ampouled under argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only