Learn More
Strontium chloride, anhydrous, 99.5% (metals basis), Thermo Scientific Chemicals
Description
Strontium chloride is widely used in fireworks for the source of red color. It is also used in dental and biological research. It is the precursor material to other compounds of strontium. It is used to prepare yellow strontium chromate, a corrosion inhibitor for aluminum. It is also employed in glass-making, sea water aquaria, and metallurgy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 1250°C |
Melting Point | 873°C |
Physical Form | Powder |
Quantity | 100 g |
Assay Percent Range | 99.5%, metal basis |
Solubility Information | Freely soluble in water. Very slightly soluble in ethanol and acetone. |
Formula Weight | 158.53 |
Refractive Index | 1.65 |
Sensitivity | Hygroscopic |
Density | 3.052 |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
W06K054 | Certificate of Analysis | 23/09/2024 | 016790.22,016790.14,016790.36 |
U06K027 | Certificate of Analysis | 20/08/2024 | 016790.22,016790.14,016790.36 |
S05K049 | Certificate of Analysis | 01/07/2024 | 016790.22,016790.14,016790.36 |
U15J027 | Certificate of Analysis | 14/09/2023 | 016790.22,016790.14,016790.36 |
D20U031 | Certificate of Analysis | 07/12/2021 | 016790.22,016790.14,016790.36 |
Safety and Handling

- Strontium chloride, anhydrous
- Danger
- Serious eye damage/eye irritation Category 1
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Strontium chloride
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.