missing translation for 'onlineSavingsMsg'
Learn More
Strontium chloride, anhydrous, 99.5% (metals basis), Thermo Scientific Chemicals Product Code.: 11430490

Strontium chloride, anhydrous, 99.5% (metals basis), Thermo Scientific Chemicals

Product Code. 11430490
500 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11430490

Brand: Thermo Scientific Alfa Aesar 016790.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Strontium chloride is widely used in fireworks for the source of red color. It is also used in dental and biological research. It is the precursor material to other compounds of strontium. It is used to prepare yellow strontium chromate, a corrosion inhibitor for aluminum. It is also employed in glass-making, sea water aquaria, and metallurgy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10476-85-4
Molecular Formula Cl2Sr
MDL Number MFCD00011249

Specifications

Boiling Point 1250°C
Melting Point 873°C
Physical Form Powder
Quantity 500 g
Assay Percent Range 99.5%, metal basis
Solubility Information Freely soluble in water. Very slightly soluble in ethanol and acetone.
Formula Weight 158.53
Refractive Index 1.65
Sensitivity Hygroscopic
Density 3.052
Chemical Name or Material Strontium Chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW06K054 Certificate TypeCertificate of Analysis Date23/09/2024 Catalog Number016790.22,016790.14,016790.36
Lot NumberU06K027 Certificate TypeCertificate of Analysis Date20/08/2024 Catalog Number016790.22,016790.14,016790.36
Lot NumberS05K049 Certificate TypeCertificate of Analysis Date01/07/2024 Catalog Number016790.22,016790.14,016790.36
Lot NumberU15J027 Certificate TypeCertificate of Analysis Date14/09/2023 Catalog Number016790.22,016790.14,016790.36
Lot NumberL27Z009 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number016790.22,016790.14,016790.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Strontium chloride, anhydrous, 99.5% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.