missing translation for 'onlineSavingsMsg'
Learn More

Strontium fluoride, 99% min, Thermo Scientific Chemicals

£56.88 - £669.33

Chemical Identifiers

CAS 7783-48-4
Molecular Formula F2Sr
Molecular Weight (g/mol) 125.62
MDL Number MFCD00011251
InChI Key FVRNDBHWWSPNOM-UHFFFAOYSA-L
Synonym strontium difluoride, unii-efy8gjs81z, strontium fluoride srf2, efy8gjs81z, strontium 2+ ion difluoride, fluoride, sr, fluoride, strontium 2+ difluoride, beta-bromo isovaleric acid, ksc379a4r, strontium fluoride 250g
PubChem CID 82210
IUPAC Name strontium;difluoride
SMILES [F-].[F-].[Sr++]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11316608
View Documents
Thermo Scientific Alfa Aesar
012201.30
250 g
£56.88
250g
Please to purchase this item. Need a web account? Register with us today!
11336608
View Documents
Thermo Scientific Alfa Aesar
012201.A1
1 kg
£162.81
1kg
Please to purchase this item. Need a web account? Register with us today!
11326608
View Documents
Thermo Scientific Alfa Aesar
012201.A7
5 kg
£669.33
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Strontium fluoride is used as an optical material for a small range of special applications. It also used as use is as a carrier of strontium-90 radioisotope in radioisotope thermoelectric generators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7783-48-4
125.62
FVRNDBHWWSPNOM-UHFFFAOYSA-L
82210
[F-].[F-].[Sr++]
F2Sr
MFCD00011251
strontium difluoride, unii-efy8gjs81z, strontium fluoride srf2, efy8gjs81z, strontium 2+ ion difluoride, fluoride, sr, fluoride, strontium 2+ difluoride, beta-bromo isovaleric acid, ksc379a4r, strontium fluoride 250g
strontium;difluoride

Specifications

1473°C
White
F2Sr
14,8842
Slightly soluble in water (11;7mg at 18°C/100mL), dilute acids; Decomposes in strong acids
[F-].[F-].[Sr++]
125.62
125.62
Odorless
4.240 g/mL
7783-48-4
Powder
MFCD00011251
strontium difluoride, unii-efy8gjs81z, strontium fluoride srf2, efy8gjs81z, strontium 2+ ion difluoride, fluoride, sr, fluoride, strontium 2+ difluoride, beta-bromo isovaleric acid, ksc379a4r, strontium fluoride 250g
FVRNDBHWWSPNOM-UHFFFAOYSA-L
strontium;difluoride
82210
≤99%
1.442
Strontium fluoride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 232-000-3

missing translation for 'rtecsNumber' : WK8925000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only