Learn More
Strontium nitrate, Puratronic™, 99.9965% (metals basis), Thermo Scientific Chemicals
Description
Strontium nitrate is used in the production of road flares and in fireworks. It serves as a colorant (red) in pyrotechnics and is also used as an oxidizer in pyrotechnics. It mixed with glycolic acid and used to reduce the sensation of skin irritation. It is also used in electrophysiology experiments.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 570°C |
Physical Form | Powder |
Quantity | 25 g |
Assay Percent Range | 99.9965% (metals basis) |
Solubility Information | Soluble in water and ammonia. Slightly soluble in alcohol,ethanol and acetone. Insoluble in nitric acid. |
IUPAC Name | strontium(2+) dinitrate |
Formula Weight | 211.63 |
Sensitivity | Hygroscopic |
Density | 2.99 g/mL |
Chemical Name or Material | Strontium nitrate |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
P31H052 | Certificate of Analysis | 24/10/2023 | 010879.06,010879.22,010879.88,010879.14 |
S13I036 | Certificate of Analysis | 27/06/2022 | 010879.06,010879.22,010879.14 |
Y02H020 | Certificate of Analysis | 12/04/2022 | 010879.06,010879.22,010879.14 |
21744 | Certificate of Analysis | 12/11/2021 | 010879.06,010879.22,010879.14 |
T11F007 | Certificate of Analysis | 12/11/2021 | 010879.06,010879.22,010879.14 |
Safety and Handling


- Strontium nitrate
- Danger
- Serious eye damage/eye irritation Category 1
- Oxidising solid Category 1
- H271-May cause fire or explosion; strong oxidiser.
- H318-Causes serious eye damage.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P310-Immediately call a POISON CENTER/doctor/.
- P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
- MIXTURE LIST-Contains : Strontium nitrate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.