missing translation for 'onlineSavingsMsg'
Learn More
Strontium nitrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals Product Code.: 11334898

Strontium nitrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11334898
25 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11334898

Brand: Thermo Scientific Alfa Aesar 043193.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used in the manufacture of lights, flares, flame tube, the TV picture tube with the red light emitting agent, red fireworks, optical glass and phosphor. It is used as a colorant (red) in pyrotechnics and is also used as an oxidizer in pyrotechnics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10042-76-9
Molecular Formula N2O6Sr
Molecular Weight (g/mol) 211.63
MDL Number MFCD00011248
InChI Key DHEQXMRUPNDRPG-UHFFFAOYSA-N
Synonym strontium nitrate, strontium dinitrate, nitric acid, strontium salt, strontium ii nitrate 1:2, unii-bdg873aqzl, nitrate de strontium french, hsdb 787, strontium nitrate sr no3 2, bdg873aqzl, nitric acid, strontium salt 2:1Show MoreShow Less
PubChem CID 24848
IUPAC Name strontium(2+) dinitrate
SMILES [Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 570°C
Physical Form Crystalline
Quantity 25 g
Assay Percent Range 99.999% (metals basis)
Solubility Information Soluble in water,liquid ammonia. Slightly soluble in ethanol and acetone. Insoluble in nitric acid.
Formula Weight 211.63
Sensitivity Hygroscopic
Density 2.99
Chemical Name or Material Strontium nitrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN24L017 Certificate TypeCertificate of Analysis Date26/02/2025 Catalog Number043193.22,043193.14
Lot NumberX03I062 Certificate TypeCertificate of Analysis Date13/10/2022 Catalog Number043193.22,043193.14
Lot NumberW01I024 Certificate TypeCertificate of Analysis Date15/09/2022 Catalog Number043193.22,043193.14
Lot NumberM25I033 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number043193.22,043193.14
Lot NumberP28I014 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number043193.22,043193.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Strontium nitrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.