missing translation for 'onlineSavingsMsg'
Learn More

Strontium sulfide, 99.9% (metals basis), Thermo Scientific Chemicals

£93.20 - £452.00

Chemical Identifiers

CAS 1314-96-1
Molecular Formula SSr
Molecular Weight (g/mol) 119.68
MDL Number MFCD00049560
InChI Key ZEGFMFQPWDMMEP-UHFFFAOYSA-N
Synonym strontium sulfide, strontium monosulfide, strontium sulphide, strontiumsulfid, hsdb 12
PubChem CID 14820
IUPAC Name strontium(2+) sulfanediide
SMILES [S--].[Sr++]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11483850
View Documents Promotion Details
Thermo Scientific Alfa Aesar
089895.09
10 g
£93.20
10g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11473850
View Documents Promotion Details
Thermo Scientific Alfa Aesar
089895.18
50 g
£452.00
50g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Strontium sulfide is used in chemical research and as organic intermediates. It is also used as depilatory. It is also used as Isotope labelled enantiomer of Voriconazole (V760000), an antifungal agent and an ergosterol biosynthesis inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1314-96-1
119.68
ZEGFMFQPWDMMEP-UHFFFAOYSA-N
14820
[S--].[Sr++]
SSr
MFCD00049560
strontium sulfide, strontium monosulfide, strontium sulphide, strontiumsulfid, hsdb 12
strontium(2+) sulfanediide

Specifications

>2000°C
Gray
SSr
14,8849
Slightly soluble in water; Decomposes in acids
[S--].[Sr++]
119.68
119.68
Like rotten eggs
3.70 g/mL
Strontium sulfide
1314-96-1
Powder
MFCD00049560
strontium sulfide, strontium monosulfide, strontium sulphide, strontiumsulfid, hsdb 12
ZEGFMFQPWDMMEP-UHFFFAOYSA-N
strontium(2+) sulfanediide
14820
99.9%
Hygroscopic
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P234-P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c

H290-H301-H314

missing translation for 'einecsNumber' : 215-249-2

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only