missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ Stuart™ Pressmatic Glass PTFE Ceramic Dispensers
Sets precise volume using a robust, easy to use, self-locking rotating ring. Stuart™ Pressmatic Glass PTFE Ceramic Dispensers deliver priming reagent straight back into the bottle using a recirculation valve to make use safer and less wasteful.
Brand: Cole-Parmer™ 10112-81
This item is not returnable.
View return policy
Description
The Pressmatic dispensers have an innovative design that maximizes safety and ease of use. A recirculation valve means that reagent used for priming is delivered straight back into the bottle making it safer and less wasteful. Once the dispenser has been fitted to the bottle, it is fully rotational for optimum positioning. The precise volume setting is by a self-locking rotating ring that is robust, easy to use and will not move during operation. The piston is manufactured from high-grade ceramic eliminating abrasion and sticking. The liquid pathway is made up of glass, PTFE and ceramic to give outstanding chemical resistance. The whole unit can be autoclaved without dismantling. Each Pressmatic dispenser is supplied with 3 x bottle adapters to fit most types of vessel and a works calibrated certificate. A full range of spare parts are available.- New ergonomic design with recirculation valve
- Chemically resistant and non-sticking ceramic piston
- Simple and accurate volume setting
- Fully rotational on bottle for optimum positioning
- Accuracy within 0.7%.
- Supplied with calibration certificate
- Can be autoclaved at 121°C
Specifications
<0.7% | |
Autoclavable | |
5 to 30 mL | |
0.50 mL |
Bottletop Dispenser | |
Glass, PTFE, Ceramic | |
Bottle |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cole-Parmer™ Stuart™ Pressmatic Glass PTFE Ceramic Dispensers > Capacity: 5 to 30mL
Spot an opportunity for improvement?Share a Content Correction