Learn More
Techne™ Stuart™ Skirted 96-well PCR Plates
These 96-well PCR plates are skirted to reduce plate distortion and provide stability.
Brand:  Techne™ 16210-31
 View more versions of this product
Product Code. 13058523
  • £183.00 / Pack of 25
Estimated Shipment: 16-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Fully skirted to reduce distortion and promote stability
  • Available in Black, Clear and White
Specifications
Specifications
Standard
White
25
25 Pk.
8 x 12
96
Low
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Techne™ Stuart™ Skirted 96-well PCR Plates > Stuart™ Skirted 96-well PCR Plates, Color: White